Skip to Content
Merck
All Photos(1)

Documents

262986

Sigma-Aldrich

Ytterbium

chips, 99.9% trace rare earth metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Yb
CAS Number:
Molecular Weight:
173.04
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.23

Assay

99.9% trace rare earth metals basis

form

chips

reaction suitability

reagent type: catalyst
core: ytterbium

resistivity

28 μΩ-cm, 20°C

bp

1196 °C (lit.)

mp

824 °C (lit.)

density

6.54 g/mL at 25 °C (lit.)

SMILES string

[Yb]

InChI

1S/Yb

InChI key

NAWDYIZEMPQZHO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

When reacted with aromatic aldimines, followed by oxidants, produces vicinal diimine compounds.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Tao Zhang et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 19(2), 739-748 (2012-11-21)
Based on a donor-acceptor framework, several conjugates have been designed and prepared in which an electron-donor moiety, ytterbium(III) porphyrinate (YbPor), was linked through an ethynyl bridge to an electron-acceptor moiety, boron dipyrromethene (BODIPY). Photoluminescence studies demonstrated efficient energy transfer from
S Rydberg et al.
Optics express, 21(6), 6681-6688 (2013-04-03)
In this work we present experimental evidence that the valence instability of the ytterbium ion play a key role for the observed photodarkening mechanism in Yb-doped fiber lasers. Luminescence and excitation spectroscopy performed on UV irradiated Yb/Al doped silica glass
Jianhua Wang et al.
Optics express, 20(27), 28373-28378 (2012-12-25)
A 100 W-class all-fiber linearly-polarized single-mode fiber laser at 1120 nm with an optical efficiency of 50% was demonstrated. The laser consists of a 4.2 m long Yb-doped polarization maintaining fiber with a core diameter of 10 μm and a
Zhen Liu et al.
Biomaterials, 34(30), 7444-7452 (2013-07-16)
Nanoparticulate contrast agents have attracted a great deal of attention along with the rapid development of modern medicine. Here, a binary contrast agent based on PAA modified BaYbF5:Tm nanoparticles for direct visualization of gastrointestinal (GI) tract has been designed and
Jin, W-S. et al.
Chemical Communications (Cambridge, England), 1101-1101 (1998)

Articles

A significant limiting factor for wearable electronics and wireless sensors is the finite amount of energy that can be stored in on-board batteries.

Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service