Skip to Content
Merck
All Photos(1)

Key Documents

249998

Sigma-Aldrich

Titanium(III) chloride solution

~10 wt. % in 20-30 wt. % hydrochloric acid

Synonym(s):

Titanium trichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
TiCl3
CAS Number:
Molecular Weight:
154.23
MDL number:
UNSPSC Code:
12352301
PubChem Substance ID:

Pricing and availability is not currently available.

reaction suitability

reagent type: catalyst
core: titanium

concentration

~10 wt. % in 20-30 wt. % hydrochloric acid

density

1.192 g/mL at 25 °C

SMILES string

Cl[Ti](Cl)Cl

InChI

1S/3ClH.Ti/h3*1H;/q;;;+3/p-3

InChI key

YONPGGFAJWQGJC-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Catalyst for:
Hydrogen desorption properties[1]
Nitrogen-nitrogen reductive bond cleavage[2]

Reagent for:
Photocatalyst synthesis[3]
Preparation of nanostructures titania/cetyltrimethyammonium bromide nanoskeleton[4]

Other Notes

Exact concentration stated on label

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S. Isobe, et al.,
Applied Physics Letters, 99 (2011)
Shuxi Dai et al.
Nanoscale research letters, 5(11), 1829-1835 (2010-12-03)
Highly crystalline TiO(2) nanostructures were prepared through a facile inorganic acid-assisted hydrothermal treatment of hexagonal-structured assemblies of nanocrystalline titiania templated by cetyltrimethylammonium bromide (Hex-ncTiO(2)/CTAB Nanoskeleton) as starting materials. All samples were characterized by X-ray diffraction (XRD) and transmission electron microscopy
D. Meroni, et al
Catalysis Today, 161, 169-174 (2011)
Catalytic oxidation of hydrazo derivatives promoted by a TiCl3/HBr system.
Eyal Drug et al.
Journal of the American Chemical Society, 129(45), 13784-13785 (2007-10-26)
Yan Zhang et al.
Organic & biomolecular chemistry, 9(13), 4977-4982 (2011-05-21)
N-N bond cleavage in hydrazines is widely used in the preparation of amines and thus occupies a significant place in organic synthesis. In this paper, we report a new method for the reductive cleavage of N-N bonds in hydrazines by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service