Skip to Content
Merck
All Photos(1)

Key Documents

203408

Sigma-Aldrich

Holmium(III) chloride hexahydrate

99.999%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HoCl3 · 6H2O
CAS Number:
Molecular Weight:
379.38
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

Assay

99.999%

reaction suitability

reagent type: catalyst
core: holmium

mp

165 °C (dec.) (lit.)

SMILES string

[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].Cl[Ho](Cl)Cl

InChI

1S/3ClH.Ho.6H2O/h3*1H;;6*1H2/q;;;+3;;;;;;/p-3

InChI key

BUQFCXABQYNXLP-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chun-Lin Lin et al.
BMC medical genomics, 13(1), 69-69 (2020-05-16)
Chromothripsis is an event of genomic instability leading to complex chromosomal alterations in cancer. Frequent long-range chromatin interactions between transcription factors (TFs) and targets may promote extensive translocations and copy-number alterations in proximal contact regions through inappropriate DNA stitching. Although

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service