Skip to Content

2025 - 1月及農曆春節預計貨物安排,請參考此頁訊息

Merck
All Photos(1)

Key Documents

ADVH14DBC133

AChemBlock

2-(2-Methoxyphenyl)propan-2-amine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
201.7
MDL number:

Pricing and availability is not currently available.

Iupac Name

2-(2-methoxyphenyl)-2-propanamine hydrochloride

Inchi Code

1S/C10H15NO.ClH/c1-10(2,11)8-6-4-5-7-9(8)12-3;/h4-7H,11H2,1-3H3;1H

InChI key

YHQHTXFCLGHENC-UHFFFAOYSA-N

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
GF10253179GF96137837GF78358438
铜 foil, 25mm disks, thickness 0.075mm, half hard, 99.95+%

GF03625051

铜 foil, 25mm disks, thickness 0.7mm, half hard, 99.95+%

GF10253179

铜 foil, 25mm disks, thickness 1.0mm, half hard, 99.95+%

GF96137837

铜 foil, 25mm disks, thickness 0.25mm, half hard, 99.95+%

GF78358438

assay

99.95%

assay

≥99.95%

assay

≥99.95%

assay

99.95%

form

foil

form

foil

form

foil

form

foil

density

8.94 g/mL at 25 °C (lit.)

density

8.94 g/mL at 25 °C (lit.)

density

8.94 g/mL at 25 °C (lit.)

density

8.94 g/mL at 25 °C (lit.)

bp

2567 °C (lit.)

bp

2567 °C (lit.)

bp

2567 °C (lit.)

bp

2567 °C (lit.)

mp

1083.4 °C (lit.)

mp

1083.4 °C (lit.)

mp

1083.4 °C (lit.)

mp

1083.4 °C (lit.)

storage temp.

Store at Room Tem.

Purity

95%

Country of Origin

CN

Product Link

https://www.achemblock.com/p41616-2-2-methoxyphenyl-propan-2-amine-hydrochloride.html

Synonym

2-(2-methoxyphenyl)propan-2-amine hydrochloride

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://www.achemblock.com/p41616-2-2-methoxyphenyl-propan-2-amine-hydrochloride.html


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service