Skip to Content
Merck
All Photos(1)

Key Documents

AABH99A4C2C2

AA BLOCKS, INC.

N-Cyclopropyl-3-methyl-3-azetidinamine 2HCl

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
126.2
MDL number:

Iupac Name

N-cyclopropyl-3-methyl-3-azetidinamine

Inchi Code

1S/C7H14N2/c1-7(4-8-5-7)9-6-2-3-6/h6,8-9H,2-5H2,1H3

InChI key

QWZGBZNTIUPYQV-UHFFFAOYSA-N

storage temp.

2~8C

Purity

97%

Country of Origin

CN

Product Link

https://www.aablocks.com/prod/149724-22-1

Synonym

N-Cyclopropyl-3-methyl-3-azetidinamine 2HCl

MSDS

https://www.aablocks.com/downloadSds?cas=149724-22-1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service