Fire Up Your Research
Neuroinflammation Biomarker Immunoassays
Neuroimmunology & Neuroinflammation Immunoassays
The Role of Immunoassays in Neuroimmunology and Neuroinflammation Research
Neuroinflammation plays a key role in the chronic processes of neurodegenerative diseases resulting from traumatic brain injury (TBI) and spinal cord injury. Inflammation is associated with disease progression in Alzheimer’s disease (AD), Parkinson’s disease, amyotrophic lateral sclerosis (ALS), and multiple sclerosis (MS). Elevated pro-inflammatory markers in the aging brain can increase the risk for cognitive impairment and the development of neurodegenerative disease.
We offer MILLIPLEX® multiplex assays for the Luminex® instrument platform and high sensitivity SMC® assays for the SMCxPRO® instrument platform to meet the needs of neuroscience researchers. These immunoassay tools enable scientists to explore neuroimmunology and neuroinflammation by monitoring biomarkers in advance of neurodegenerative disease onset, during chronic disease, and during potential therapeutic treatments to slow disease progression.
Figure 1.Concentration of MILLIPLEX® Human Panel A analytes in neurodegenerative disease samples. CSF samples were compared between healthy controls (n=9), mild cognitive impairment (MCI) samples (n=10), and AD samples (n=13). **p < 0.005 ***p > 0.0005.
Cytokine Analysis in Neurodegenerative Disease
Quantitating cytokine and chemokine markers of neuroinflammation and biomarkers of neurodegeneration in cerebral spinal fluid (CSF), serum/plasma, cell culture, and tissue lysate samples allow researchers to study disease progression associated with neuroinflammation and neurodegeneration. Neuroinflammation may play a role in AD and other neurodegenerative diseases, highlighting the utility of broadly profiling cytokines in CSF samples. The MILLIPLEX® Human Cytokine/Chemokine/Growth Factor Panel A and Panel B enable multiplexing of many types of bioassays reducing time, labor, and costs over traditional methods.
Ultrasensitive Analysis of Neurodegenerative Disease Biomarkers
The need for blood-based biomarkers of AD has driven a continuous search for novel candidates. However, the identification of blood-based biomarkers may be limited by the sensitivity of standard immunoassay methodologies. High sensitivity immunoassay technologies, such as SMC® technology, can transform neurodegenerative disease research by enabling the measurement of low-abundant proteins in a variety of biofluids and create opportunities for the identification of novel biomarkers.
Figure 2.CSF measurement of AD biomarkers. Analysis of control and AD CSF samples with each SMC® kit (SMC® Phospho-Tau (T181) Kit, SMC® Total Tau Kit, SMC® TDP-43 Kit, SMC® SNAP-25 Kit) revealed statistically significant differences in phosphorylated Tau T181 (p<0.02), total Tau (p<0.03), TDP-43 (p<0.01), and SNAP-25 (p<0.02).
Request Info on Neuroinflammation Immunoassay Products
For Research Use Only. Not For Use In Diagnostic Procedures.
Related Product Resources
- High Sensitivity Immunoassays in Neuroscience
Neuroscience biomarker research expands with high-sensitivity immunoassays for low-abundant biomarkers.
- Multiplex Immunoassay for Alzheimer’s Biomarkers
MILLIPLEX® multiplex immunoassays detect Alzheimer's biomarkers in neuroscience research using high sensitivity technology.
- Alzheimer’s Biomarker Detection | MILLIPLEX® Assays
Multiplex detection of Alzheimer's biomarkers with MILLIPLEX® Human Amyloid Beta and Tau Magnetic Bead Panel aids research.
- Neuroscience Multiplex Assays | MILLIPLEX® Assays
Neuroscience multiplex assays measure biomarkers to understand neurodegenerative diseases, disorders, and neuropeptide signaling.
- /language-masters/en/technical-documents/product-supporting/smc-technologies/measuring-endogenous-inflammation-biomarkers
- Power of High Sensitivity Immunology Assays | SMC® Technology
Use the power of high sensitivity immunology assays with SMC® technology to precisely characterize low-abundant cytokines like those from Th1/Th2 and Th17 cells.
- Immunology Multiplex Assays | MILLIPLEX®
Immunology multiplex assays like MILLIPLEX® allow simultaneous investigation of immune biomarkers, saving time and resources in research.
- Multiplex Inflammation Signaling | MILLIPLEX® Assays
MILLIPLEX® assays bridge immunology and cell signaling for inflammation studies, including T cell signaling and sepsis signaling.
- Overview of Luminex® Multiplex Assay Technology | MILLIPLEX® Multiplex Assays
Discover the benefits of MILLIPLEX® multiplex assays, based on Luminex® xMAP® multiplex assay technology, that provide consistent, high-quality results and see how these multiplex biomarker Luminex® assays are being used to advance research.
- What Is SMC® Technology? | Ultrasensitive Immunoassay Technology
Explore ultrasensitive immunoassay technology with SMC® technology, including its biomarker detection platform, advantages of high sensitivity assays, and more.
- Conferma® ELISAs: Designed for Sample Detection & Lot Consistency
Learn about the Conferma® ELISA development and manufacturing methods that provide strong sample detection and long-term assay and lot consistency, giving you confidence in your research.
- Selecting Immunoassay Platforms
Guide on selecting immunoassay platforms including multiplex vs. singleplex assays, instruments, software, and services.