US EPA Method TO-10/IP-8 and ASTM® D4861: GC Analysis of Pesticides on SPB®-608 after Collection/Desorption using ORBO™-1000
CONDITIONS
sample/matrix
polyurethane foam (PUF) spiked with 250 ng each pesticide
adsorbent tube
ORBO-1000 (20557)
column
SPB-608, 30 m × 0.53 mm I.D., 0.5 μm film (25312)
oven
150 °C (1 min) to 270 °C at 4 °C/min
detector
ECD
carrier gas
helium, 7 mL/min
injection
1 μL
Description
Legal Information
ASTM is a registered trademark of American Society for Testing and Materials
ORBO is a trademark of Sigma-Aldrich Co. LLC
SPB is a registered trademark of Merck KGaA, Darmstadt, Germany