GC Analysis of 2,5-Dimethoxytetrahydrofuran Enantiomers on Astec® CHIRALDEX™ G-TA
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Astec CHIRALDEX G-TA, 30 m x 0.25 mm I.D., 0.12 μm (73033AST)
oven
70 °C
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
helium, 30 psi
injection
0.2 μL, 80:1 split
sample
2 mg/mL in ethanol
peaks 1 & 2: 2,5-dimethoxytetrahydrofuran enantiomers
peak 3: meso-isomer
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIRALDEX is a trademark of Sigma-Aldrich Co. LLC