Merck
Ruthenium catalysed cross metathesis with fluorinated olefins.
Chemical communications (Cambridge, England) (2002-09-21)
S Imhof, S Randl, S Blechert
ABSTRACT
The E-selective cross metathesis (CM) of fluorinated olefins with various functionalised alkenes in good to excellent yields is reported.
Product Description
Hoveyda-Grubbs Catalyst® M720 ChemBeads
Pricing and availability is not currently available.
Sigma-Aldrich
Hoveyda-Grubbs Catalyst® M720, Umicore, 97%
Pricing and availability is not currently available.