Merck
  • Monoacylglycerol lipase inhibitor JZL184 prevents HIV-1 gp120-induced synapse loss by altering endocannabinoid signaling.
Monoacylglycerol lipase inhibitor JZL184 prevents HIV-1 gp120-induced synapse loss by altering endocannabinoid signaling.
Neuropharmacology (2017-10-25)
Xinwen Zhang, Stanley A Thayer
ABSTRACT
Monoacylglycerol lipase (MGL) hydrolyzes 2-arachidonoylglycerol to arachidonic acid and glycerol. Inhibition of MGL may attenuate neuroinflammation by enhancing endocannabinoid signaling and decreasing prostaglandin (PG) production. Almost half of HIV infected individuals are afflicted with HIV-associated neurocognitive disorder (HAND), a neuroinflammatory disease in which cognitive decline correlates with synapse loss. HIV infected cells shed the envelope protein gp120 which is a potent neurotoxin that induces synapse loss. Here, we tested whether inhibition of MGL, using the selective inhibitor JZL184, would prevent synapse loss induced by gp120. The number of synapses between rat hippocampal neurons in culture was quantified by imaging clusters of a GFP-tagged antibody-like protein that selectively binds to the postsynaptic scaffolding protein, PSD95. JZL184 completely blocked gp120-induced synapse loss. Inhibition of MGL decreased gp120-induced interleukin-1β (IL-1β) production and subsequent potentiation of NMDA receptor-mediated calcium influx. JZL184-mediated protection of synapses was reversed by a selective cannabinoid type 2 receptor (CB
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
LY 320135, ≥98% (HPLC)
Sigma-Aldrich
Arachidonoyl-AMC, fatty acid amide hydrolase substrate
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
AH6809, ≥98%, crystalline solid or supercooled liquid
Sigma-Aldrich
AM630, ≥90% (HPLC)