- High-throughput printing via microvascular multinozzle arrays.
High-throughput printing via microvascular multinozzle arrays.
Advanced materials (Deerfield Beach, Fla.) (2012-10-31)
Christopher J Hansen, Rajat Saksena, David B Kolesky, John J Vericella, Stephen J Kranz, Gregory P Muldowney, Kenneth T Christensen, Jennifer A Lewis
PMID23109104
ABSTRACT
Microvascular multinozzle arrays are designed and fabricated for high-throughput printing of functional materials. Ink-flow uniformity within these multigeneration, bifurcating microchannel arrays is characterized by computer modeling and microscopic particle image velocimetry (micro-PIV) measurements. Both single and dual multinozzle printheads are produced to enable rapid printing of multilayered periodic structures over large areas (≈1 m(2)).
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol) diacrylate, average Mn ~5,800
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol) dimethacrylate, average Mn ~12,500
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol) diacrylate, average Mn ~14,600
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol) diacrylate, average Mn ~12,500