Merck
  • Evaluation of bi-functionalized mesoporous silicas as reversed phase/cation-exchange mixed-mode sorbents for multi-residue solid phase extraction of veterinary drug residues in meat samples.
Evaluation of bi-functionalized mesoporous silicas as reversed phase/cation-exchange mixed-mode sorbents for multi-residue solid phase extraction of veterinary drug residues in meat samples.
Talanta (2017-02-06)
Natalia Casado, Damián Pérez-Quintanilla, Sonia Morante-Zarcero, Isabel Sierra
ABSTRACT
A SBA-15 type mesoporous silica was synthesized and bi-functionalized with octadecylsilane (C18) or octylsilane (C8), and sulfonic acid (SO
Product Description
Supelco
Cimaterol, VETRANAL®, analytical standard
Supelco
Tulobuterol hydrochloride, VETRANAL®, analytical standard
Sigma-Aldrich
Chloro(dimethyl)octylsilane, 97%
Supelco
Carazolol, analytical standard
Supelco
Flunixin, VETRANAL®, analytical standard
Supelco
Clenproperol, VETRANAL®, analytical standard
Supelco
Brombuterol hydrochloride, VETRANAL®, analytical standard
Supelco
Mabuterol hydrochloride, VETRANAL®, analytical standard
Sigma-Aldrich
(3-Mercaptopropyl)triethoxysilane, ≥80% (GC), technical