Merck
  • Inhibitory Effects of Garcinia cambogia Extract on CYP2B6 Enzyme Activity.
Inhibitory Effects of Garcinia cambogia Extract on CYP2B6 Enzyme Activity.
Planta medica (2017-03-14)
Jun Sang Yu, Min Sun Choi, Jong Suk Park, Shaheed Ur Rehman, Katsunori Nakamura, Hye Hyun Yoo
ABSTRACT
This study assessed the inhibitory effects of
Sigma-Aldrich
4-Hydroxymephenytoin, ≥98% (HPLC)
Sigma-Aldrich
Furafylline, ≥98% (HPLC)
Supelco
(−)-Calcium hydroxycitrate tribasic, analytical standard
Sigma-Aldrich
Dextrorphan