Merck
  • Single sensor for multiple analytes in different optical channel: Applying for multi-ion response modulation.
Single sensor for multiple analytes in different optical channel: Applying for multi-ion response modulation.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2017-04-30)
Chunshuang Liang, Shimei Jiang
ABSTRACT
A Schiff-base, (2,4-di-tert-butyl-6-((2-hydroxyphenyl-imino)-methyl)phenol) (L), has been improved to function as a simultaneous multi-ion probe in different optical channel. The probe changes from colorless to orangish upon being deprotonated by F
Sigma-Aldrich
3,5-Di-tert-butyl-2-hydroxybenzaldehyde, 99%