Merck
  • Developmental attenuation of N-methyl-D-aspartate receptor subunit expression by microRNAs.
Developmental attenuation of N-methyl-D-aspartate receptor subunit expression by microRNAs.
Neural development (2015-09-19)
Caroline Corbel, Israel Hernandez, Bian Wu, Kenneth S Kosik
ABSTRACT
N-methyl-D-aspartate receptors (NMDARs) are a subtype of ionotropic glutamate receptors and are expressed throughout the central nervous system (CNS). Their activity is required for excitatory synaptic transmission, the developmental refinement of neural circuits and for the expression of many forms of synaptic plasticity. NMDARs are obligate heterotetramers and the expression of their constituent subunits is developmentally and anatomically regulated. In rodent cortex and hippocampus, the GluN2B subunit is expressed at high levels early in development and decreases to plateau levels later while expression of the GluN2A subunit has a concomitant increase. Regulation of GluN2A and GluN2B expressions are incompletely understood. Here, we showed the influence of miRNAs in this process. Two miRNAs, miR-19a and miR-539 can influence the levels of NMDARs subunits, as they target the mRNAs encoding GluN2A and GluN2B respectively. MiR-539 also modified the expression of the transcription factor REST, a known regulator of NMDAR subunit expression. miR-19a and miR-539, in collaboration with REST, serve to set the levels of GluN2A and GluN2B precisely during development. These miRNAs offer an entry point for interventions that affect plasticity and a novel approach to treat neurodegenerative diseases.
Sigma-Aldrich
Anti-NMDAR1 Antibody, clone 54.1, clone 54.1, Chemicon®, from mouse
Sigma-Aldrich
Anti-NR2B Antibody, clone BWJHL, clone BWJHL, Upstate®, from mouse
Sigma-Aldrich
Anti-NMDAR2A Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid