Merck
  • Bacterial E3 Ubiquitin Ligase IpaH4.5 of Shigella flexneri Targets TBK1 To Dampen the Host Antibacterial Response.
Bacterial E3 Ubiquitin Ligase IpaH4.5 of Shigella flexneri Targets TBK1 To Dampen the Host Antibacterial Response.
Journal of immunology (Baltimore, Md. : 1950) (2015-12-25)
Zirui Zheng, Congwen Wei, Kai Guan, Yuan Yuan, Yanhong Zhang, Shengli Ma, Ye Cao, Fang Wang, Hui Zhong, Xiang He
ABSTRACT
IFN regulatory factors play a pivotal role in many cellular processes, including inflammatory and immune responses. Their activation is tightly regulated by TANK-binding kinase 1 (TBK1). In response to microbial components, TBK1 activates IFN regulatory factor 3 (IRF3) and cytokine expression. In this article, we show that TBK1 is a novel target of the IpaH4.5 protein, a Shigella type III effector possessing E3 ubiquitin ligase activity. Remarkably, IpaH4.5 interacts with TBK1 and promotes its K48-linked polyubiquitylation. Consequently, polyubiquitylated TBK1 undergoes proteasome-dependent degradation, which perturbs the phosphorylation, nuclear translocation, and activation of IRF3. Because IRF3 and TBK1 are required for restricting Shigella growth, we propose that the polyubiquitylation and degradation of TBK1 during Shigella infection are new bacterial strategies to modulate the host antibacterial responses.
Sigma-Aldrich
Anti-c-Myc−Peroxidase antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-HA antibody produced in mouse, clone HA-7, ascites fluid
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone B-5-1-2, purified from hybridoma cell culture