Merck
  • Disposable screen printed sensor for the electrochemical detection of delta-9-tetrahydrocannabinol in undiluted saliva.
Disposable screen printed sensor for the electrochemical detection of delta-9-tetrahydrocannabinol in undiluted saliva.
Chemistry Central journal (2016-01-26)
Ceri Wanklyn, Dan Burton, Emma Enston, Carrie-Ann Bartlett, Sarah Taylor, Aleksandra Raniczkowska, Murdo Black, Lindy Murphy
ABSTRACT
Cannabis has an adverse effect on the ability to drive safely, therefore a rapid disposable test for Δ(9)-tetrahydrocannabinol (Δ(9)-THC), the psychoactive component of cannabis, is highly desirable for roadside testing. A screen printed carbon electrode is used for the N-(4-amino-3-methoxyphenyl)-methanesulfonamide mediated detection of Δ(9)-THC in saliva. Mediator placed in an overlayer was galvanostatically oxidized and reacted with Δ(9)-THC to give an electrochemically active adduct which could be detected by chronoamperometric reduction. Detection of 25-50 ng/mL Δ(9)-THC spiked into undiluted saliva was achieved with a response time of 30 s. A trial of the sensors with four cannabis smokers showed sensitivity of 28 %, specificity of 99 % and accuracy of 52 %. Rapid electrochemical detection of Δ(9)-THC in undiluted saliva has been demonstrated using a disposable sensor, however the sensitivity is lower than acceptable. Further optimization of the assay and sensor format is required to improve the sensitivity of response to Δ(9)-THC.
Product Description
Supelco
(−)-trans-Δ9-THC solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Pricing and availability is not currently available.