Merck
  • Experimental and Computational Evidence for the Reduction Mechanisms of Aromatic N-oxides by Aqueous Fe(II)-Tiron Complex.
Experimental and Computational Evidence for the Reduction Mechanisms of Aromatic N-oxides by Aqueous Fe(II)-Tiron Complex.
Environmental science & technology (2015-12-05)
Yiling Chen, Hao Dong, Huichun Zhang
ABSTRACT
A combined experimental-theoretical approach was taken to elucidate the reduction mechanisms of five representative aromatic N-oxides (ANOs) by Fe(II)-tiron complex and to identify the rate-limiting step. Based on the possible types of complexes formed with the reductant, three groups of ANOs were studied: type I refers to those forming 5-membered ring complexes through the N and O atoms on the side chain; type II refers to those forming 6-membered ring complexes through the N-oxide O atom and the O atom on the side chain; and type III refers to complexation through the N-oxide O atom only. Density functional theory calculations suggested that the elementary reactions, including protonation, N-O bond cleavage, and the second electron transfer processes, are barrierless, indicating that the first electron transfer is rate-limiting. Consistent with the theoretical results, the experimental solvent isotope effect, KIEH, for the reduction of quinoline N-oxide (a type III ANO) was obtained to be 1.072 ± 0.025, suggesting protonation was not involved in the rate-limiting step. The measured nitrogen kinetic isotope effect, KIEN, for the reduction of pyridine N-oxide (a type III ANO) (1.022 ± 0.006) is in good agreement with the calculated KIEN for its first electron transfer (1.011-1.028), confirming that the first electron transfer is rate-limiting. Electrochemical cell experiments demonstrated that the electron transfer process can be facilitated significantly by type I complexation with FeL2(6-) (1:2 Fe(II)-tiron complex), to some extent by type II complexation with free Fe(II), but not by weak type III complexation.
Sigma-Aldrich
Pyridine N-oxide, 95%
Sigma-Aldrich
Pyridine-15N, 98 atom % 15N