Merck
  • Identification of Novel Biomarker and Therapeutic Target Candidates for Diagnosis and Treatment of Follicular Adenoma.
Identification of Novel Biomarker and Therapeutic Target Candidates for Diagnosis and Treatment of Follicular Adenoma.
Cancer genomics & proteomics (2015-11-07)
Xianyin Lai, Shaoxiong Chen
ABSTRACT
Follicular adenoma is a type of benign and encapsulated nodule in the thyroid gland, but some adenomas have the potential to progress to follicular carcinoma. Therefore, it is important to monitor the state and progress of follicular adenoma in the clinic and discover drug development targets for the treatment of follicular adenoma to prevent its worsening to follicular carcinoma. Currently, the study of biomarkers and therapeutic targets lacks applications of up-to-date technologies, including proteomics and bioinformatics. To discover novel protein biomarker and therapeutic target candidates, a liquid chromatography-tandem mass spectrometry approach was applied to directly compare follicular adenoma with normal thyroid tissue samples. The proteomics analysis revealed 114 protein biomarker candidates out of 1,780 identified and quantified proteins. A comprehensive approach to prioritize the biomarker candidates by category and rank revealed CD63, DDB1, TYMP, VDAC2, and DCXR as the top five biomarker candidates. Upstream regulator analysis using the Ingenuity Pathway Analysis (IPA) software discovered four therapeutic target candidates for follicular adenoma, including TGFB1, MYC, ANGPT2, and NFE2L2. This study provided biomarker and therapeutic target candidates for a follow-up study, which will facilitate monitoring and treatment of follicular adenoma.
Product Description
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Urea, BioUltra, for molecular biology, 99% (T)
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Ammonium bicarbonate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea, meets USP testing specifications
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Acetonitrile, for preparative HPLC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)