Merck
A novel role for Plk4 in regulating cell spreading and motility.
Oncogene (2014-09-02)
C O Rosario, K Kazazian, F S W Zih, O Brashavitskaya, Y Haffani, R S Z Xu, A George, J W Dennis, C J Swallow
ABSTRACT
Polo family kinase 4 (Plk4) is required for mitotic progression, and is haploinsufficient for tumor suppression and timely hepatocyte polarization in regenerating liver. At the same time, recent evidence suggests that Plk4 expression may have a role in clinical cancer progression, although the mechanisms are not clear. Here we identify a gene expression pattern predictive of reduced motility in Plk4(+/-) murine embryonic fibroblasts (MEFs) and validate this prediction with functional assays of cell spreading, migration and invasion. Increased Plk4 expression enhances cell spreading in Plk4(+/-) MEFs and migration in human embryonic kidney 293T cells, and increases invasion by DLD-1 colon cancer cells. Plk4 depletion impairs invasion of wild-type MEFs and suppresses invasion by MDA-MB231 breast cancer cells. Cytoskeletal reorganization and development of polarity are impaired in Plk4-deficient cells that have been stimulated to migrate. Endogenous Plk4 phosphorylated at the autophosphorylation site S305 localizes to the protrusions of motile cells, coincident with the RhoA GEF Ect2, GTP-bound RhoA and the RhoA effector mDia. Taken together, our findings reveal an unexpected activity of Plk4 that promotes cell migration and may underlie an association between increased Plk4 expression, cancer progression and death from metastasis in solid tumor patients.
Sigma-Aldrich
Anti-Vinculin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-γ-Tubulin antibody, Mouse monoclonal, clone GTU-88, ascites fluid
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Plk4
Sigma-Aldrich
MISSION® esiRNA, targeting human PLK4
Sigma-Aldrich
MISSION® pLKO.1-puro Luciferase shRNA Control Plasmid DNA, shRNA sequence targeting luciferase
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O