Merck
  • TAT-ODD-p53 enhances the radiosensitivity of hypoxic breast cancer cells by inhibiting Parkin-mediated mitophagy.
TAT-ODD-p53 enhances the radiosensitivity of hypoxic breast cancer cells by inhibiting Parkin-mediated mitophagy.
Oncotarget (2015-05-31)
Rong Zheng, Qiwei Yao, Guozhu Xie, Shasha Du, Chen Ren, Yuxia Wang, Yawei Yuan
ABSTRACT
Radiation therapy has an important role in the treatment of breast cancer. Dysfunction p53 and hypoxia are typical biological characteristics of breast cancer that constitute barriers to the efficacy of radiotherapy. Mitophagy plays a protective role in cellular homeostasis under hypoxic conditions, while mitophagy is inhibited by p53 in normal cells. We explored the effects of a p53 fusion protein, TAT-ODD-p53, on the radiosensitivity of hypoxic breast cancer cells both in vitro and in vivo, as well as investigating the related molecular mechanisms. We found that selective accumulation of TAT-ODD-p53 occurred under hypoxic conditions and significantly increased tumor cell radiosensitivity both in vitro and in vivo. Mitophagy had an important role in maintaining hypoxia-induced radioresistance. Mitophagy was inhibited by TAT-ODD-p53 and this inhibition was suppressed by over-expression of Parkin in hypoxic irradiated breast cancer cells. In addition, mitophagy was induced by deletion of p53, with this effect being weakened by Parkin knockdown at a low oxygen tension. By interacting with Parkin, p53 inhibited the translocation of Parkin to the mitochondria, disrupting the protective mitophagy process. These results suggest that TAT-ODD-p53 has a significant and preferential radiosensitizing effect on hypoxic breast cancer cells by inhibition of Parkin-mediated mitophagy.
Sigma-Aldrich
MISSION® esiRNA, targeting human ATG7
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Atg7
Sigma-Aldrich
Ethopropazine hydrochloride, ≥98% (HPLC), powder
Sigma-Aldrich
MISSION® esiRNA, targeting human TP53
Roche
In Situ Cell Death Detection Kit, TMR red, sufficient for ≤50 tests
Sigma-Aldrich
DAPI, for nucleic acid staining