Merck
  • Fibroblast activation protein alpha expression identifies activated fibroblasts after myocardial infarction.
Fibroblast activation protein alpha expression identifies activated fibroblasts after myocardial infarction.
Journal of molecular and cellular cardiology (2015-09-01)
Jochen Tillmanns, Daniel Hoffmann, Yasmin Habbaba, Jan D Schmitto, Daniel Sedding, Daniela Fraccarollo, Paolo Galuppo, Johann Bauersachs
ABSTRACT
Fibroblast activation protein α (FAP) is a membrane-bound serine protease expressed by activated fibroblasts during wound healing in the skin. Expression of FAP after myocardial infarction (MI) and potential effects on cardiac wound healing are largely unknown. MI was induced in rats and FAP expression was analyzed at 3, 7 and 28 days post-MI by microarray, Western blot and immunohistochemistry. In human hearts after MI, a FAP(+) fibroblast population was identified, and characterized by immunohistochemistry for prolyl-4-hydroxylase β, α-smooth muscle actin, Thy-1 and vimentin. Signaling pathways leading to FAP expression were studied in human cardiac fibroblasts by Western blot and ELISA using TGFβ1, TGF-beta type I-receptor (TGFbR1)-inhibitor SB431542 or the MAPK-inhibitor U0126 as well as siRNA targeting SMAD2 and SMAD3. Finally, fibroblasts were assayed for FAP-dependent migration (modified Boyden-chamber), proliferation (BrdU-assay) and gelatinolytic activity by gelatin zymography. In rats, FAP expression was increased after MI especially in the peri-infarct area peaking at 7 days post-MI. Co-localization analysis identified the majority of FAP(+) cells as activated proto-myofibroblasts and myofibroblasts. Concordantly, FAP(+) fibroblasts were abundant in ischemic tissue of human hearts after MI, but not in healthy control hearts. In vitro, FAP was induced by TGFβ1 via the canonical SMAD2/SMAD3 pathway. Depletion of FAP in fibroblasts reduced migratory capacity, while proliferation was not affected. Gelatin zymography revealed gelatinase activity by fibroblast-derived FAP. In this study, we show for the first time the expression of FAP in activated fibroblasts after MI and its activation by TGFβ1. Effects of FAP on fibroblast migration and gelatinolytic activity indicate a potential role in cardiac wound healing and remodeling.
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
SAFC
HEPES
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
SAFC
HEPES
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
MISSION® esiRNA, targeting human GLMN
Sigma-Aldrich
MISSION® esiRNA, targeting human FAP
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Glmn