Merck
  • (99m)Tc-amitrole as a novel selective imaging probe for solid tumor: In silico and preclinical pharmacological study.
(99m)Tc-amitrole as a novel selective imaging probe for solid tumor: In silico and preclinical pharmacological study.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences (2015-05-10)
B M Essa, T M Sakr, Mohammed A Khedr, F A El-Essawy, A A El-Mohty
ABSTRACT
Lactoperoxidase (LPO) inhibitors are very selective for solid tumor due to their high binding affinity to the LPO enzyme. A computational study was used to select top-ranked LPO inhibitor (alone and in complex with (99m)Tc) with high in silico affinity. The novel prepared (99m)Tc-amitrole complex demonstrated both in silico and in vivo high affinity toward solid tumors.(99m)Tc-amitrole was radio-synthesized with a high radiochemical yield (89.7±3.25). It showed in vitro stability for up to 6h. Its preclinical evaluation in solid tumor-bearing mice showed high retention and biological accumulation in solid tumor cells with a high Target/Non-Target (T/NT) ratio equal to 4.9 at 60min post-injection. The data described previously could recommend (99m)Tc-amitrole as potential targeting scintigraphic probe for solid tumor imaging.
Sigma-Aldrich
3-Amino-1,2,4-triazole, ≥95% (TLC)
Sigma-Aldrich
4H-1,2,4-Triazol-3-amine, AldrichCPR
Sigma-Aldrich
Sodium iodide, 99.999% trace metals basis
Sigma-Aldrich
Sodium iodide, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium iodide, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium iodide, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%