Merck
  • Catalytic behaviour in the ring-opening polymerisation of organoaluminiums supported by bulky heteroscorpionate ligands.
Catalytic behaviour in the ring-opening polymerisation of organoaluminiums supported by bulky heteroscorpionate ligands.
Dalton transactions (Cambridge, England : 2003) (2014-12-24)
Jose A Castro-Osma, Carlos Alonso-Moreno, Agustín Lara-Sánchez, Antonio Otero, Juan Fernández-Baeza, Luis F Sánchez-Barba, Ana M Rodríguez
ABSTRACT
A series of alkyl organoaluminium complexes based on bulky heteroscorpionate ligands were designed as catalysts for the ring-opening polymerisation of cyclic esters. Thus, the treatment of AlX3 (X = Me, Et) with bulky acetamide or thioacetamide heteroscorpionate ligands nbptamH (1) [nbptamH = N-naphthyl-2,2-bis(3,5-dimethylpyrazol-1-yl)thioacetamide], fbpamH (2) [fbpamH = N-fluorenyl-2,2-bis(3,5-dimethylpyrazol-1-yl)acetamide], ptbptamH (3) [ptbptamH = N-phenyl-2,2-bis(3,5-di-tert-butylpyrazol-1-yl)thioacetamide], ntbptamH (4) [ntbptamH = N-naphthyl-2,2-bis(3,5-di-tert-butylpyrazol-1-yl)thioacetamide], ptbpamH (5) [ptbpamH = N-phenyl-2,2-bis(3,5-di-tert-butylpyrazol-1-yl)acetamide] and (S)-mtbpamH (6) [(S)-mtbpamH = (S)-(−)-N-α-methylbenzyl-2,2-bis(3,5-di-tert-butylpyrazol-1-yl)acetamide] for 1 hour at 0 °C afforded the dialkyl aluminium complexes [AlX2{κ(2)-nbptam}] (X = Me 7, Et 8), [AlX2{κ(2)-fbpam}] (X = Me 9, Et 10), [AlX2{κ(2)-ptbptam}] (X = Me 11, Et 12), [AlX2{κ(2)-ntbptam}] (X = Me 13, Et 14), [AlX2{κ2(-)ptbpam}] (X = Me 15, Et 16) and [AlX2{κ(2)-(S)-mtbpam}] (X = Me 17, Et 18). The structures of the complexes were determined by spectroscopic methods and the X-ray crystal structure of 14 was also established. The alkyl-containing aluminium complexes 7–18 can act as efficient single-component initiators for the ring-opening polymerisation of ε-caprolactone and rac-lactide. The polymerisations are living, as evidenced by the narrow polydispersities of the isolated polymers and the linear nature of the number average molecular weight versus conversion plot. Finally, a comparative study of ring-opening polymerisation for new bulky heteroscorpionate aluminium initiators and the less congested aluminium analogues is reported.
Sigma-Aldrich
Phenyl isothiocyanate, 99%, for protein sequencing
Sigma-Aldrich
1-Naphthyl isothiocyanate, 95%
Sigma-Aldrich
Phenyl isothiocyanate, reagent grade, 98%
Sigma-Aldrich
Phenyl isothiocyanate, for HPLC derivatization, the detection of alcohols and amines, ≥99.0%
Sigma-Aldrich
Phenyl isothiocyanate, Sigma Grade, 8.36 M, suitable for solid phase protein sequencing analysis, ≥99% (GC), liquid
Sigma-Aldrich
trans-2-[3-(4-tert-Butylphenyl)-2-methyl-2-propenylidene]malononitrile, ≥98%
Sigma-Aldrich
Sodium iodide, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium iodide, 99.999% trace metals basis
Sigma-Aldrich
3,6-Dimethyl-1,4-dioxane-2,5-dione, 99%
Sigma-Aldrich
Sodium iodide, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium iodide, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%