Merck
  • Identification of the expressed protein and the impact of change in ascorbate peroxidase activity related to endodormancy breaking in Pyrus pyrifolia.
Identification of the expressed protein and the impact of change in ascorbate peroxidase activity related to endodormancy breaking in Pyrus pyrifolia.
Plant physiology and biochemistry : PPB (2014-12-02)
Yoshihiro Takemura, Katsuou Kuroki, Mingfeng Jiang, Kazuhiro Matsumoto, Fumio Tamura
ABSTRACT
Endodormancy is an important feature of perennial deciduous fruit trees that survive in the extreme climates brought about by seasonal variation. To acquire a comprehensive knowledge of the biochemical processes occurring just before endodormancy breaking, the buds collected in the pre-breaking period (PP) phase were used as samples to identify the proteins related to the breaking of endodormancy in the Japanese pear (Pyrus pyrifolia Nakai). Using nano-ESI-LC-MS/MS analysis, 96 proteins were overlapped by analyses of three times and identified as expressed proteins at the PP stage. Among these proteins, dehydrin, several classes of heat shock proteins (HSP), auxin-binding protein, and auxin-induced protein were identified in the floral bud in the PP stage. The majority of these proteins were involved primarily in the oxidation-reduction process. We focused on catalase (CAT), peroxidase (POD), and ascorbate peroxidase (APX) as enzymes regulating the levels of hydrogen peroxide (H2O2) in the bud. From measurements taken during the deepest period (DP), PP, mid-breaking period (MP), and late-breaking period (LP) of endodormancy, CAT activity decreased gradually, while APX activity also decreased from DP to MP, but then increased rapidly during LP. Protein data for PP and the rapid increase in APX activity observed in LP provided knowledge of the biochemical processes that regulate the consecutive transition from endodormancy breaking to ecodormancy induction in the Japanese pear.
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Supelco
Potassium hydroxide concentrate, 0.1 M KOH in water (0.1N), Eluent concentrate for IC
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Potassium hydroxide, tested according to Ph. Eur.
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Potassium hydroxide, anhydrous, ≥99.95% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Potassium hydroxide, BioXtra, ≥85% KOH basis
Supelco
Potassium hydroxide solution, volumetric, 8.0 M KOH (8.0N)
Sigma-Aldrich
Potassium hydroxide, ≥85% KOH basis, pellets, white
Sigma-Aldrich
Potassium hydroxide, technical, ≥85%, powder
Sigma-Aldrich
Potassium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, 85-100.5%, pellets
Sigma-Aldrich
Potassium hydroxide, pellets, reag. Ph. Eur., ≥85%
Sigma-Aldrich
Potassium hydroxide, ACS reagent, ≥85%, pellets
Sigma-Aldrich
Potassium hydroxide, semiconductor grade, pellets, 99.99% trace metals basis (Purity excludes sodium content.)
Sigma-Aldrich
Potassium hydroxide, reagent grade, 90%, flakes
Sigma-Aldrich
Potassium hydroxide, puriss. p.a., ≥86% (T), pellets
Sigma-Aldrich
Potassium hydroxide solution, 45 wt. % in H2O