Merck
  • A pragmatic definition of therapeutic synergy suitable for clinically relevant in vitro multicompound analyses.
A pragmatic definition of therapeutic synergy suitable for clinically relevant in vitro multicompound analyses.
Molecular cancer therapeutics (2014-04-24)
Muhammad Kashif, Claes Andersson, Magnus Åberg, Peter Nygren, Tobias Sjöblom, Ulf Hammerling, Rolf Larsson, Mats G Gustafsson
ABSTRACT
For decades, the standard procedure when screening for candidate anticancer drug combinations has been to search for synergy, defined as any positive deviation from trivial cases like when the drugs are regarded as diluted versions of each other (Loewe additivity), independent actions (Bliss independence), or no interaction terms in a response surface model (no interaction). Here, we show that this kind of conventional synergy analysis may be completely misleading when the goal is to detect if there is a promising in vitro therapeutic window. Motivated by this result, and the fact that a drug combination offering a promising therapeutic window seldom is interesting if one of its constituent drugs can provide the same window alone, the largely overlooked concept of therapeutic synergy (TS) is reintroduced. In vitro TS is said to occur when the largest therapeutic window obtained by the best drug combination cannot be achieved by any single drug within the concentration range studied. Using this definition of TS, we introduce a procedure that enables its use in modern massively parallel experiments supported by a statistical omnibus test for TS designed to avoid the multiple testing problem. Finally, we suggest how one may perform TS analysis, via computational predictions of the reference cell responses, when only the target cell responses are available. In conclusion, the conventional error-prone search for promising drug combinations may be improved by replacing conventional (toxicology-rooted) synergy analysis with an analysis focused on (clinically motivated) TS.
Sigma-Aldrich
Fluorouracil, meets USP testing specifications
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder
Supelco
5-Fluorouracil, analytical standard
Fluorouracil, European Pharmacopoeia (EP) Reference Standard
USP
Fluorouracil, United States Pharmacopeia (USP) Reference Standard
Supelco
Fluorouracil, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Oxaliplatin, powder
Supelco
Oxaliplatin, Pharmaceutical Secondary Standard; Certified Reference Material
Oxaliplatin, European Pharmacopoeia (EP) Reference Standard
USP
Oxaliplatin, United States Pharmacopeia (USP) Reference Standard