Merck
  • A BODIPY-luminol chemiluminescent resonance energy-transfer (CRET) cassette for imaging of cellular superoxide.
A BODIPY-luminol chemiluminescent resonance energy-transfer (CRET) cassette for imaging of cellular superoxide.
Organic & biomolecular chemistry (2014-12-17)
S Bag, J-C Tseng, J Rochford
ABSTRACT
Spectroscopic and in cellulo studies are here reported on the very first BODIPY-luminol chemiluminescent resonance energy-transfer (CRET) cassette where the luminol CL agent is covalently linked to the BODIPY energy-transfer acceptor in a molecular dyad. The efficiency of intramolecular CRET investigated for the BODIPY-luminol dyad was found to be 64% resulting in a dual emissive response. Successful in cellulo biochemiluminescence via CRET was achieved in PMA activated splenocytes.
Sigma-Aldrich
p-Toluenesulfonic acid monohydrate, ≥99% (calc. to H2O free subst.)
Sigma-Aldrich
Luminol, 97%
Sigma-Aldrich
Sodium borohydride, granular, 99.99% trace metals basis
Sigma-Aldrich
Sodium borohydride, ReagentPlus®, 99%
Sigma-Aldrich
p-Toluenesulfonic acid monohydrate, ACS reagent, ≥98.5%
Sigma-Aldrich
Sodium borohydride, purum p.a., ≥96% (gas-volumetric)
Sigma-Aldrich
Sodium borohydride, caplets (18 × 10 × 8 mm), 98%
Sigma-Aldrich
p-Toluenesulfonic acid monohydrate, ReagentPlus®, ≥98%
Sigma-Aldrich
Sodium borohydride, powder, ≥98.0%
Sigma-Aldrich
Sodium borohydride, granular, 10-40 mesh, 98%
Sigma-Aldrich
Luminol, ≥97% (HPLC)
Sigma-Aldrich
VenPure® SF, powder
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Sodium borohydride solution, 2.0 M in triethylene glycol dimethyl ether
Sigma-Aldrich
2,3-Dichloro-5,6-dicyano-p-benzoquinone, 98%
Sigma-Aldrich
Boron, crystalline, 1 cm, 99.7% trace metals basis
Sigma-Aldrich
Triethylamine, purum, ≥99% (GC)
Supelco
Tetrahydrofuran, analytical standard
Supelco
Tetrahydrofuran, HPLC grade, ≥99.9%, inhibitor-free
Sigma-Aldrich
Sodium borohydride solution, ~12 wt. % in 14 M NaOH
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Sigma-Aldrich
Boron, ≥95% (boron), amorphous powder
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, for protein sequence analysis, ampule, ≥99.5% (GC)
Supelco
Tetrahydrofuran, Selectophore, ≥99.5%
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)