Merck
  • Chimeric antigen receptors against CD33/CD123 antigens efficiently target primary acute myeloid leukemia cells in vivo.
Chimeric antigen receptors against CD33/CD123 antigens efficiently target primary acute myeloid leukemia cells in vivo.
Leukemia (2014-02-08)
I Pizzitola, F Anjos-Afonso, K Rouault-Pierre, F Lassailly, S Tettamanti, O Spinelli, A Biondi, E Biagi, D Bonnet
ABSTRACT
As significant numbers of acute myeloid leukemia (AML) patients are still refractory to conventional therapies or experience relapse, immunotherapy using T cells expressing chimeric antigen receptors (CARs) might represent a valid treatment option. AML cells frequently overexpress the myeloid antigens CD33 and CD123, for which specific CARs can be generated. However, CD33 is also expressed on normal hematopoietic stem/progenitor cells (HSPCs), and its targeting could potentially impair normal hematopoiesis. In contrast, CD123 is widely expressed by AML, while low expression is detected on HSPCs, making it a much more attractive target. In this study we describe the in vivo efficacy and safety of using cytokine-induced killer (CIK) cells genetically modified to express anti-CD33 or anti-CD123 CAR to target AML. We show that both these modified T cells are very efficient in reducing leukemia burden in vivo, but only the anti-CD123 CAR has limited killing on normal HSPCs, thus making it a very attractive immunotherapeutic tool for AML treatment.
Sigma-Aldrich
D-Luciferin, synthetic
Sigma-Aldrich
D-Luciferin, synthetic, BioXtra, ≥99% (HPLC)