Merck
  • Development and validation of a rapid immunochromatographic assay for detection of Middle East respiratory syndrome coronavirus antigen in dromedary camels.
Development and validation of a rapid immunochromatographic assay for detection of Middle East respiratory syndrome coronavirus antigen in dromedary camels.
Journal of clinical microbiology (2015-01-30)
Daesub Song, Gunwoo Ha, Wissam Serhan, Yassir Eltahir, Mohammed Yusof, Farouq Hashem, Elsaeid Elsayed, Bahaaeldin Marzoug, Assem Abdelazim, Salama Al Muhairi
ABSTRACT
We present here a rapid immunochromatographic assay for the detection of Middle East respiratory syndrome coronavirus (MERS-CoV) antigen in the nasal swabs of dromedary camels. The assay is based on the detection of MERS-CoV nucleocapsid protein in a short time frame using highly selective monoclonal antibodies at room temperature. The relative sensitivity and specificity of the assay were found to be 93.90% and 100%, respectively, compared to that of the UpE and open reading frame 1A (Orf1A) real-time reverse transcriptase PCR (RT-PCR). The results suggest that the assay developed here is a useful tool for the rapid diagnosis and epidemiological surveillance of MERS-CoV infection in dromedary camels.
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Imidazole, ≥99% (titration), crystalline
Sigma-Aldrich
Imidazole, for molecular biology, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Imidazole, ACS reagent, ≥99% (titration)
Sigma-Aldrich
TWEEN® 20, viscous liquid
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Imidazole, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Imidazole, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Imidazole, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Imidazole, European Pharmacopoeia (EP) Reference Standard
Millipore
Urea solution, suitable for microbiology, 40% in H2O
SAFC
Sodium chloride solution, 5 M