Merck
  • Plant development. Integration of growth and patterning during vascular tissue formation in Arabidopsis.
Plant development. Integration of growth and patterning during vascular tissue formation in Arabidopsis.
Science (New York, N.Y.) (2014-08-12)
Bert De Rybel, Milad Adibi, Alice S Breda, Jos R Wendrich, Margot E Smit, Ondřej Novák, Nobutoshi Yamaguchi, Saiko Yoshida, Gert Van Isterdael, Joakim Palovaara, Bart Nijsse, Mark V Boekschoten, Guido Hooiveld, Tom Beeckman, Doris Wagner, Karin Ljung, Christian Fleck, Dolf Weijers
ABSTRACT
Coordination of cell division and pattern formation is central to tissue and organ development, particularly in plants where walls prevent cell migration. Auxin and cytokinin are both critical for division and patterning, but it is unknown how these hormones converge upon tissue development. We identify a genetic network that reinforces an early embryonic bias in auxin distribution to create a local, nonresponding cytokinin source within the root vascular tissue. Experimental and theoretical evidence shows that these cells act as a tissue organizer by positioning the domain of oriented cell divisions. We further demonstrate that the auxin-cytokinin interaction acts as a spatial incoherent feed-forward loop, which is essential to generate distinct hormonal response zones, thus establishing a stable pattern within a growing vascular tissue.
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Pricing and availability is not currently available.
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Pricing and availability is not currently available.
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Pricing and availability is not currently available.
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Pricing and availability is not currently available.
Sigma-Aldrich
Magnesium chloride, BioReagent, suitable for insect cell culture, ≥97.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Pricing and availability is not currently available.
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Pricing and availability is not currently available.