Merck
  • BIM upregulation and ROS-dependent necroptosis mediate the antitumor effects of the HDACi Givinostat and Sorafenib in Hodgkin lymphoma cell line xenografts.
BIM upregulation and ROS-dependent necroptosis mediate the antitumor effects of the HDACi Givinostat and Sorafenib in Hodgkin lymphoma cell line xenografts.
Leukemia (2014-02-25)
S L Locatelli, L Cleris, G G Stirparo, S Tartari, E Saba, M Pierdominici, W Malorni, A Carbone, A Anichini, C Carlo-Stella
ABSTRACT
Relapsed/refractory Hodgkin's lymphoma (HL) is an unmet medical need requiring new therapeutic options. Interactions between the histone deacetylase inhibitor Givinostat and the RAF/MEK/ERK inhibitor Sorafenib were examined in HDLM-2 and L-540 HL cell lines. Exposure to Givinostat/Sorafenib induced a synergistic inhibition of cell growth (range, 70-80%) and a marked increase in cell death (up to 96%) due to increased H3 and H4 acetylation and strong mitochondrial injury. Gene expression profiling indicated that the synergistic effects of Givinostat/Sorafenib treatment are associated with the modulation of cell cycle and cell death pathways. Exposure to Givinostat/Sorafenib resulted in sustained production of reactive oxygen species (ROS) and activation of necroptotic cell death. The necroptosis inhibitor Necrostatin-1 prevented Givinostat/Sorafenib-induced ROS production, mitochondrial injury, activation of BH3-only protein BIM and cell death. Knockdown experiments identified BIM as a key signaling molecule that mediates Givinostat/Sorafenib-induced oxidative death of HL cells. Furthermore, in vivo xenograft studies demonstrated a 50% reduction in tumor burden (P<0.0001), a 5- to 15-fold increase in BIM expression (P < 0.0001) and a fourfold increase in tumor necrosis in Givinostat/Sorafenib-treated animals compared with mice that received single agents. These results provide a rationale for exploring Givinostat/Sorafenib combination in relapsed/refractory HL.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Bcl2l11
Sigma-Aldrich
Tetramethylrhodamine ethyl ester perchlorate, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Hematoxylin, certified by the Biological Stain Commission
Sigma-Aldrich
Hematoxylin