Merck
  • The quetiapine active metabolite N-desalkylquetiapine and the neurotensin NTS₁ receptor agonist PD149163 exhibit antidepressant-like effects on operant responding in male rats.
The quetiapine active metabolite N-desalkylquetiapine and the neurotensin NTS₁ receptor agonist PD149163 exhibit antidepressant-like effects on operant responding in male rats.
Experimental and clinical psychopharmacology (2014-10-07)
Todd M Hillhouse, Zachary Shankland, Katelin S Matazel, Ashley A Keiser, Adam J Prus
ABSTRACT
Major depressive disorder is the most common mood disorder in the United States and European Union; however, the limitations of clinically available antidepressant drugs have led researchers to pursue novel pharmacological treatments. Clinical studies have reported that monotherapy with the atypical antipsychotic drug quetiapine produces a rapid reduction in depressive symptoms that is apparent after 1 week of treatment, and it is possible that the active metabolite N-desalkylquetiapine, which structurally resembles an antidepressant drug, produces antidepressant effects. Neuropharmacological evaluations of the neurotensin NTS1 receptor agonist PD149163 suggest antidepressant efficacy, but the effects of a NTS₁ receptor agonist in an antidepressant animal model have yet to be reported. The present study examined the antidepressant-like effects of N-desalkylquetiapine, PD14916, quetiapine, the tricyclic antidepressant drug imipramine, the atypical antipsychotic drug risperidone, and the typical antipsychotic drug raclopride on responding in male Sprague-Dawley rats trained on a differential-reinforcement-of-low-rate 72-s operant schedule, a procedure used for screening antidepressant drugs. Quetiapine, PD149163, risperidone, and imipramine exhibited antidepressant-like effects by increasing the number of reinforcers earned, decreasing the number of responses emitted, and shifting the interresponse time (IRT) distributions to the right. N-Desalkylquetiapine produced a partial antidepressant-like effect by decreasing the number of responses emitted and producing a rightward shift in the IRT distributions, but it did not significantly alter the number of reinforcers earned. Raclopride decreased reinforcers and responses. These data suggest that N-desalkylquetiapine likely contributes to quetiapine's antidepressant efficacy and identify NTS₁ receptor activation as a potential novel pharmacologic strategy for antidepressant drugs.
Sigma-Aldrich
Risperidone, ≥98% (HPLC), powder
Sigma-Aldrich
Sodium trichloroacetate, 97%
USP
Risperidone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
PD 149163 tetrahydrochloride hydrate, ≥90% (HPLC)
Risperidone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Neurotensin, ≥90% (HPLC)
Risperidone for system suitability, European Pharmacopoeia (EP) Reference Standard