Merck
  • Application of bispecific antibody against antigen and hapten for immunodetection and immunopurification.
Application of bispecific antibody against antigen and hapten for immunodetection and immunopurification.
Experimental & molecular medicine (2013-09-28)
Hyori Kim, Sunyoung Park, Hwa Kyoung Lee, Junho Chung
ABSTRACT
We present a bispecific antibody that recognizes an antigen and a hapten and can be applied to various biological assays, including immunoblotting and immunoprecipitation. In immunoblot analysis of serum, an anti-C5 × anti-cotinine bispecific tandem single-chain variable fragment (scFv)-Fc fusion protein and cotinine-conjugated horseradish peroxidase (HRP) generated a clean signal without the high background that was observed in a parallel experiment using HRP-conjugated goat anti-rabbit immunoglobulin G (Fc-specific) antibody. In immunoprecipitation analysis of serum, use of the bispecific tandem scFv-Fc fusion protein and cotinine-crosslinked magnetic beads significantly reduced the amount of protein contaminants compared with a parallel experiment done with protein A agarose beads. In subsequent immunoblot analysis, use of cotinine-HRP as the secondary probe instead of HRP-conjugated goat anti-rabbit IgG (Fc-specific) antibody successfully eliminated the band corresponding to the bispecific tandem scFv-Fc fusion protein.
Sigma-Aldrich
trans-4-Cotininecarboxylic acid, 97%
Pricing and availability is not currently available.