Merck
Eugenol: a natural compound with versatile pharmacological actions.
Natural product communications (2011-02-09)
Kannissery Pramod, Shahid H Ansari, Javed Ali
ABSTRACT
Eugenol, the major constituent of clove oil, has been widely used for its anesthetic and analgesic action in dentistry. Eugenol exhibits pharmacological effects on almost all systems and our aim is to review the research work that has identified these pharmacological actions. Eugenol possesses significant antioxidant, anti-inflammatory and cardiovascular properties, in addition to analgesic and local anesthetic activity. The metabolism and pharmacokinetics of the compound in humans have been studied. Eugenol has also been used as a penetration enhancer. The compound is a very promising candidate for versatile applications, and the design of new drugs based on the pharmacological effects of eugenol could be beneficial.
Product Description
Supelco
Eugenol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Eugenol, ≥98%, FCC, FG
Supelco
Eugenol, PESTANAL®, analytical standard
Sigma-Aldrich
Eugenol, natural, ≥98%, FG
Sigma-Aldrich
Eugenol, ReagentPlus®, 99%
Supelco
Eugenol, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Eugenol, European Pharmacopoeia (EP) Reference Standard