Merck
Implementation of a mandatory donor RHD screening in Switzerland.
Transfusion and apheresis science : official journal of the World Apheresis Association : official journal of the European Society for Haemapheresis (2014-04-01)
Sofia Lejon Crottet, Christine Henny, Stefan Meyer, Franziska Still, Martin Stolz, Jochen Gottschalk, Kathrin Neuenschwander, Behrouz Mansouri Taleghani, Peter Gowland, Beat M Frey, Stefano Fontana, Hein Hustinx, Christoph Niederhauser, Christoph Gassner
ABSTRACT
Starting in 2013, blood donors must be tested at least using: (1) one monoclonal anti-D and one anti-CDE (alternatively full RhCcEe phenotyping), and (2) all RhD negative donors must be tested for RHD exons 5 and 10 plus one further exonic, or intronic RHD specificity, according to the guidelines of the Blood Transfusion Service of the Swiss Red Cross (BTS SRC). In 2012 an adequate stock of RHD screened donors was built. Of all 25,370 RhD negative Swiss donors tested in 2012, 20,015 tested at BTS Berne and 5355 at BTS Zürich, showed 120 (0.47%) RHD positivity. Thirty-seven (0.15%) had to be redefined as RhD positive. Routine molecular RHD screening is reliable, rapid and cost-effective and provides safer RBC units in Switzerland.
Sigma-Aldrich
REDExtract-N-Amp PCR ReadyMix for Blood, 12 mL sufficient for 1000 amplifications
Pricing and availability is not currently available.
Sigma-Aldrich
REDExtract-N-Amp Blood PCR Kit, sufficient for 10 extractions, sufficient for 10 amplifications
Pricing and availability is not currently available.
Sigma-Aldrich
REDExtract-N-Amp Blood PCR Kit, sufficient for 1000 extractions, sufficient for 1000 amplifications
Pricing and availability is not currently available.