Merck
  • Hemoglobin subunit beta interacts with the capsid protein and antagonizes the growth of classical swine fever virus.
Hemoglobin subunit beta interacts with the capsid protein and antagonizes the growth of classical swine fever virus.
Journal of virology (2013-03-15)
Dan Li, Hong Dong, Su Li, Muhammad Munir, Jianing Chen, Yuzi Luo, Yuan Sun, Lihong Liu, Hua-Ji Qiu
ABSTRACT
The capsid (C) protein of the Flaviviridae family members is involved in nucleocapsid formation and virion assembly. However, the influence of C protein-interacting partners on the outcome of pestivirus infections is poorly defined. In this study, hemoglobin subunit beta (HB) was identified as a C protein-binding protein by glutathione S-transferase pulldown and subsequent mass spectrometry analysis of PK-15 cells, which are permissive cells for classical swine fever virus (CSFV). Coimmunoprecipitation and confocal microscopy confirmed that HB interacts and colocalizes with the C protein in the cytoplasm. Silencing of HB with small interfering RNAs promoted CSFV growth and replication, whereas overexpression of HB suppressed CSFV replication and growth. Interestingly, HB was found to interact with retinoic acid-inducible gene I and increase its expression, resulting in increased production of type I interferon (IFN). However, HB was unable to suppress CSFV growth when the RIG-I pathway was blocked. Overall, our results suggest that cellular HB antagonizes CSFV growth and replication by triggering IFN signaling, and might represent a novel antiviral restriction factor. This study reports for the first time the novel role of HB in innate immunity.
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal ANTI-FLAG® M1 antibody, clone M1, purified immunoglobulin, buffered aqueous solution
Millipore
ANTI-FLAG® antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–TRITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Roche
X-tremeGENE HP DNA Transfection Reagent, High-performance polymer reagent for transfecting many cell lines