Merck
  • Synthesis, characteristics and luminescence properties of oligo(phenylenevinylene) dimers with a biphenyl linkage center.
Synthesis, characteristics and luminescence properties of oligo(phenylenevinylene) dimers with a biphenyl linkage center.
Chemical communications (Cambridge, England) (2003-09-19)
Feng He, Gang Cheng, Haiquan Zhang, Yan Zheng, Zengqi Xie, Bing Yang, Yuguang Ma, Shiyong Liu, Jiacong Shen
ABSTRACT
Building a new structual oligo(phenylenevinylene) by linking trimeric phenylenvinylene (TPV) through the phenylphenyl bond of a central phenylene ring was reported. The resultant TPV dimers exhibit weak intermolecular interactions and intense blue photoluminescence in the solid state as well as high phase transition temperatures up to 250 degrees C. Organic light-emitting devices (OLEDs) based on these materials display blue emission with low turn-on voltage (about 3 V), maximum luminance approaching 2000 cd m(-2) and efficiency up to 1.6 cd A(-1).
Sigma-Aldrich
2-Bromo-1,4-dimethylbenzene, 99%