- Experimental evidence for the formation of divalent ytterbium in the photodarkening process of Yb-doped fiber lasers.
Experimental evidence for the formation of divalent ytterbium in the photodarkening process of Yb-doped fiber lasers.
Optics express (2013-04-03)
S Rydberg, M Engholm
PMID23546049
ABSTRACT
In this work we present experimental evidence that the valence instability of the ytterbium ion play a key role for the observed photodarkening mechanism in Yb-doped fiber lasers. Luminescence and excitation spectroscopy performed on UV irradiated Yb/Al doped silica glass preforms and near-infrared diode pumped photodarkened fibers show a concentration increase of Yb(2+) ions. A concentration decrease in Yb(3+) could also be observed for the UV irradiated preform. The findings contribute to an increased understanding of the kinetic processes related to photodarkening in Yb-doped high power fiber lasers.