- Separation of morphine and its oxidation products by capillary zone electrophoresis.
Separation of morphine and its oxidation products by capillary zone electrophoresis.
Journal of pharmaceutical and biomedical analysis (2000-03-07)
B Proksa
PMID10704021
ABSTRACT
Separation of morphine and its oxidation products (10-S-hydroxymorphine, pseudomorphine and morphine N-oxide) by capillary zone electrophoresis in Tris-borate buffer in the presence of cyclodextrins was studied. Pyridoxine was used as an internal standard.
Product Description
Supelco
10-Hydroxymorphine solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®