Merck
  • Immunostimulatory effects of platinum compounds: correlation between sensitizing properties in vivo and modulation of receptor-mediated endocytosis in vitro.
Immunostimulatory effects of platinum compounds: correlation between sensitizing properties in vivo and modulation of receptor-mediated endocytosis in vitro.
International archives of allergy and immunology (1997-02-01)
H C Schuppe, J Kulig, U Kühn, U Lempertz, P Kind, J Knop, D Becker
ABSTRACT
The sensitizing properties of different complex salts of platinum were defined in vivo by means of the popliteal lymph node (PLN) assay in mice. Hexa- and tetrachloroplatinates were confirmed to be highly immunogenic, inducing vigorous primary immune responses in the draining PLN following single subcutaneous injections. Flow-cytometric analysis revealed a dramatic increase in the total number of cells expressing proliferating cell nuclear antigen. The majority of these cells were of the T helper phenotype (CD4+) reflecting the T-cell dependence of the PLN response induced by Pt salts such as Na2[PtCl6] or Na2[PtCl4]. In contrast, [Pt(NH3)4]Cl2 failed to elicit a significant increase in PLN cell proliferation when compared with saline-treated controls. The differential immunogenicity of the Pt compounds found in vivo directly correlated with their capacity to modulate mechanisms of receptor-mediated endocytosis in murine Langerhans cells in vitro. The reactivity of Na2[PtCl6] or Na2[PtCl4] resembled that of potent contact sensitizers in this endocytosis assay whereas [Pt(NH3)4]Cl2 proved to be mert. These results suggest that [Pt(NH3)4]Cl2 might be less harmful to humans than hexa- or tetrachloroplatinates. As demonstrated with Pt compounds, monitoring of direct effects of low-molecular-weight chemicals on antigen-presenting dendritic cells in vitro is able to predict their sensitizing potential in vivo.
Sigma-Aldrich
Ammonium tetrachloroplatinate(II), 99%
Sigma-Aldrich
Potassium tetrachloroplatinate(II), ≥99.9% trace metals basis
Sigma-Aldrich
Potassium tetrachloroplatinate(II), 98%
Sigma-Aldrich
Potassium tetrachloroplatinate(II), 99.99% trace metals basis