Merck
  • Limited proteolysis of Hansenula polymorpha yeast amine oxidase: isolation of a C-terminal fragment containing both a copper and quino-cofactor.
Limited proteolysis of Hansenula polymorpha yeast amine oxidase: isolation of a C-terminal fragment containing both a copper and quino-cofactor.
FEBS letters (1995-09-11)
J Plastino, J P Klinman
ABSTRACT
Limited proteolysis of recombinant Hansenula polymorpha yeast amino oxidase produces a 48 kDa fragment which corresponds to the C-terminal two-thirds of the protein. The fragment contains both TOPA (2,4,5-trihydroxyphenylalanine) and copper, as well as the histidine ligands implicated in copper binding. The fragment is proposed to be the domain responsible for cofactor production in yeast amine oxidase.
Sigma-Aldrich
6-Hydroxy-DL-DOPA, ≥98% (HPLC), powder
USP
Levadopa Related Compound A, United States Pharmacopeia (USP) Reference Standard