Merck
  • Generating tunable optical pulses over the ultrabroad range of 1.6-2.5 μm in GeO2-doped silica fibers with an Er:fiber laser source.
Generating tunable optical pulses over the ultrabroad range of 1.6-2.5 μm in GeO2-doped silica fibers with an Er:fiber laser source.
Optics express (2012-11-29)
E A Anashkina, A V Andrianov, M Yu Koptev, V M Mashinsky, S V Muravyev, A V Kim
ABSTRACT
We report generation of femtosecond optical pulses tunable in the 1.6-2.5 μm range using GeO2-doped core silica-cladding fibers. Optical solitons with a duration of 80-160 fs have been measured by the FROG technique in the 2-2.3 μm range. To the best of our knowledge, these are the longest wavelength temporally characterized solitons generated in silica-based fibers. We have also demonstrated more than octave-spanning femtosecond supercontinuum generation in the 1.0-2.6 μm range.
Sigma-Aldrich
Germanium(IV) oxide, (crystalline powder), 99.998% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Germanium(IV) oxide, powder, 99.999% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Germanium(IV) oxide, ≥99.99% trace metals basis
Pricing and availability is not currently available.