Merck
  • Novel silicificated PVAc/POSS composite nanofibrous mat via facile electrospinning technique: potential scaffold for hard tissue engineering.
Novel silicificated PVAc/POSS composite nanofibrous mat via facile electrospinning technique: potential scaffold for hard tissue engineering.
Colloids and surfaces. B, Biointerfaces (2012-10-31)
Yu-Mi Ha, Touseef Amna, Mi-Hee Kim, Hyun-Chel Kim, M Shamshi Hassan, Myung-Seob Khil
ABSTRACT
This study presents the fabrication of novel porous silicificated PVAc/POSS composite nanofibers by facile electrospinning technique and the interaction of synthesized mats with simulated body fluid (SBF). The physicochemical properties of the electrospun composites were determined by scanning electron microscopy, energy dispersive X-ray spectroscopy, transmission electron microscopy, electron probe micro-analysis, X-ray diffraction and thermogravimetry analysis. To examine the in vitro cytotoxicity, mouse myoblast C2C12 cells were treated with pristine and composite nanofibrous mats and the viability of cells was analyzed by cell counting kit-8 assay at regular time intervals. Our results indicated the enhanced nucleation and the formation of apatite-like structures at the surface of silicificated PVAc/POSS during the incubation of electrospun mats in SBF solution. Cytotoxicity experiments designated that the myoblasts could attach to the composite after being cultured. We observed in the present study that PVAc/POSS nanofibrous mat could support cell adhesion and guide the spreading behavior of myoblasts. We conclude that the new electrospun silicificated PVAc/POSS composite scaffold with unique porous morphology have excellent biocompatibility. Consequently, our investigation results showed that the as-spun porous PVAc/POSS composite nanofibrous scaffold could be a potential substrate for the proliferation and mineralization of osteoblasts, enhancing bone regeneration. The biocomposite mats represent a promising biomaterial to be exploited for various tissue engineering applications such as guided bone regeneration.
Sigma-Aldrich
Kollicoat® SR 30 D, 28.5-31.5% solids basis
Sigma-Aldrich
Poly(vinyl acetate), average Mw ~500,000 by GPC
Sigma-Aldrich
Poly(vinyl acetate), average Mw ~100,000 by GPC, beads
Poly(vinyl acetate) dispersion 30 per cent, European Pharmacopoeia (EP) Reference Standard