Merck
  • Ordered arrays of Raman nanosensors for ultrasensitive and location predictable biochemical detection.
Ordered arrays of Raman nanosensors for ultrasensitive and location predictable biochemical detection.
Advanced materials (Deerfield Beach, Fla.) (2012-08-14)
Xiaobin Xu, Kwanoh Kim, Huifeng Li, D L Fan
ABSTRACT
Surface enhanced Raman scattering (SERS) is sensitive enough for single-molecule biochemical detection, but it is extremely difficult to obtain a large number of SERS hotspots for sensitive and reproducible detection. It is even more challenging to assemble the hotspots at designated positions for location predictable sensing. Here, we report an original strategy for the synthesis, manipulation, and assembling of plasmonic nanocapsule SERS sensors for high-sensitivity biochemical detection at predictable locations.
Sigma-Aldrich
Rhodamine 6G, Dye content ~95 %
Sigma-Aldrich
Rhodamine 6G, Dye content 99 %
Sigma-Aldrich
Rhodamine 6G perchlorate, Dye content 99 %
Sigma-Aldrich
Rhodamine 6G, suitable for fluorescence, BioReagent
Supelco
Rhodamine 6G, analytical standard