Merck
  • Concise approach to benzisothiazol-3(2H)-one via copper-catalyzed tandem reaction of o-bromobenzamide and potassium thiocyanate in water.
Concise approach to benzisothiazol-3(2H)-one via copper-catalyzed tandem reaction of o-bromobenzamide and potassium thiocyanate in water.
The Journal of organic chemistry (2012-03-27)
Fei Wang, Chao Chen, Geng Deng, Chanjuan Xi
ABSTRACT
A concise approach to various benzisothiazol-3(2H)-one derivatives has been developed by copper-catalyzed the reaction of o-bromobenzamide derivatives with potassium thiocyanate (KSCN) in water. The reaction proceeds via a tandem reaction with S-C bond and S-N bond formation.
Sigma-Aldrich
Potassium thiocyanate solution, volumetric, 8 M KSCN
Sigma-Aldrich
1,2-Benzisothiazol-3(2H)-one, 97%
Sigma-Aldrich
Potassium thiocyanate, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium thiocyanate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Potassium thiocyanate, ACS reagent, 99%