Merck
  • HPLC separation of acetaminophen and its impurities using a mixed-mode reversed-phase/cation exchange stationary phase.
HPLC separation of acetaminophen and its impurities using a mixed-mode reversed-phase/cation exchange stationary phase.
Journal of chromatographic science (2012-03-13)
Octavian Călinescu, Irinel A Badea, Luminiţa Vlădescu, Viorica Meltzer, Elena Pincu
ABSTRACT
Determination of acetaminophen and its main impurities: 4-nitrophenol, 4'-chloroacetanilide, as well as 4-aminophenol and its degradation products, p-benzoquinone and hydroquinone has been developed and validated by a new high-performance liquid chromatography method. Chromatographic separation has been obtained on a Hypersil Duet C18/SCX column, using gradient elution, with a mixture of phosphate buffer (pH = 4.88) and methanol as a mobile phase. Analysis time did not exceed 14.5 min and good resolutions, peak shapes and asymmetries have resulted. The linearity of the method has been tested in the range of 5.0-60 µg/mL for acetaminophen and 0.5-6 µg/mL for the other compounds. The limits of detection and quantification have been also established to be lower than 0.1 µg/mL and 0.5 µg/mL, respectively. The method has been successfully applied for the analysis of commercial acetaminophen preparations.
Product Description
Supelco
Acetaminophen Related Compound B, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Supelco
4-Aminophenol (Acetaminophen Related Compound K) (Paracetamol Impurity K), Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Sigma-Aldrich
4′-Chloroacetanilide, 97%
Pricing and availability is not currently available.
Supelco
Acetaminophen Related Compound J, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Supelco
Acetaminophen Related Compound D, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.