Merck
  • Production of the polyclonal antibody against Sudan 3 and Immunoassay of Sudan dyes in food samples.
Production of the polyclonal antibody against Sudan 3 and Immunoassay of Sudan dyes in food samples.
Journal of agricultural and food chemistry (2012-02-15)
Yong Hua Qi, Wen Chong Shan, Yan Zheng Liu, Yu Jie Zhang, Jian Ping Wang
ABSTRACT
In this study, 4-aminophenylacetic acid was covalently coupled to aniline to synthesize an intermediate hapten and the intermediate hapten was coupled to β-naphthol to synthesize a tentative hapten of Sudan 3. The hapten was coupled to bovine serum albumin as the immunogen to produce the polyclonal antibody. The obtained antibody was highly specific to Sudan 3, Sudan 1, and Para red, but showed relative low binding ability to Sudan 2, Sudan 4, and Sudan red G. After evaluation of different coating antigens, a heterologous indirect competitive immunoassay was developed to multidetermine the six red dyes in food samples. The cross reactivities to the six analytes were in a range of 21-105%, and the limits of detection were in a range of 0.1-0.8 ng/mL depending on the compound. Intra- and interassay recoveries from the standard fortified blank samples were in a range of 74.5-96.3% with coefficients of variation lower than 15.1%.
Product Description
Supelco
Sudan III, analytical standard
Sigma-Aldrich
Sudan III, BioXtra, certified by the BSC, certified by the Biological Stain Commission
Sigma-Aldrich
Sudan III, Technical grade