Merck
  • Continuous synthesis process of hexagonal nanoplates of P6m ordered mesoporous silica.
Continuous synthesis process of hexagonal nanoplates of P6m ordered mesoporous silica.
Journal of the American Chemical Society (2011-07-28)
Jasper Jammaer, Titus S van Erp, Alexander Aerts, Christine E A Kirschhock, Johan A Martens
ABSTRACT
Hexagonally ordered mesoporous silica coined COK-12 was synthesized in a continuous process by combining streams of sodium silicate and citric acid/sodium citrate buffered solution of (ethylene oxide)(20)-(propylene oxide)(70)-(ethylene oxide)(20) triblock copolymer (Pluronic P123) from separate reservoirs. COK-12 precipitated spontaneously upon combining both streams at nearly neutral pH and ambient temperature. Stable intermediates of the COK-12 formation process could be prepared by limiting sodium silicate addition. Investigation of these intermediates using small-angle X-ray scattering revealed COK-12 formed via an assembly process departing from spherical uncharged core-shell P123-silica micelles. The sterical stabilization of these micelles decreased upon accumulation of silicate oligomers in their shell. Aggregation of the spherical micelles led to cylindrical micelles, which aligned and adopted the final hexagonal organization. This unprecedentedly fast formation of P6m ordered mesoporous silica was caused by two factors in the synthesis medium: the neutral pH favoring uncharged silicate oligomers and the high salt concentration promoting hydrophobic interactions with surfactant micelles leading to silica accumulation in the PEO shell. The easy continuous synthesis process is convenient for large-scale production. The platelet particle morphology with short and identical internal channels will be advantageous for many applications such as pore replication, nanotube or fiber growth, catalytic functionalization, drug delivery, film and sensor development, and in nano dyes as well as for investigation of pore diffusion phenomena.
Product Description
Supelco
(±)-Propylene oxide, analytical standard
Sigma-Aldrich
(±)-Propylene oxide, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
(±)-Propylene oxide, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium metasilicate pentahydrate, ≥95.0% (T)