Merck
  • Stereo-specific inhibition of acetyl- and butyryl-cholinesterases by enantiomers of cis,cis-decahydro-2-naphthyl-N-n-butylcarbamate.
Stereo-specific inhibition of acetyl- and butyryl-cholinesterases by enantiomers of cis,cis-decahydro-2-naphthyl-N-n-butylcarbamate.
Journal of biochemical and molecular toxicology (2011-06-02)
Ming-Cheng Lin, Shyh-Jei Yeh, I-Ru Chen, Gialih Lin
ABSTRACT
Enantiomers of cis,cis-decahydro-2-naphthyl-N-n-butylcarbamate show stereo-specific inhibition for acetylcholinesterase and butyrylcholinesterase. For both inhibition reaction, (2S,4aR,8aS)-cis,cis-decahydro-2-naphthyl-N-n- butylcarbamate is more potent than (2R,4aS,8aR)-cis,cis-decahydro-2-naphthyl-N-n-butylcarbamate. Optically pure (2S,4aR,8aS)-(-)- and (2R,4aS,8aR)-(+)-cis,cis-decahydro-2-naphthols are resolved by the porcine pancreatic lipase-catalyzed acetylation of decahydro-2-naphthols with vinyl acetate. Absolute configurations and the enantiomeric excess values of (2S,4aR,8aS)-(-)- and (2R,4aS,8aR)-(+)-cis,cis-decahydro-2-naphthols are determined from the (19)F NMR spectra of their Mosher's ester derivatives. We fail to resolve (2S,4aR,8aR)- and (2R,4aS,8aS)-trans,cis-decahydro-2-naphthols from the porcine pancreatic lipase-catalyzed acetylation of decahydro-2-naphthols with vinyl acetate.
Product Description
Supelco
Vinyl acetate solution, certified reference material, 5000 μg/mL in acetonitrile
Pricing and availability is not currently available.
Supelco
Vinyl acetate, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Vinyl acetate, contains 3-20 ppm hydroquinone as inhibitor, ≥99%
Pricing and availability is not currently available.
Vinyl acetate, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.