Merck
  • Structurally diverse amphiphiles exhibit biphasic modulation of GABAA receptors: similarities and differences with neurosteroid actions.
Structurally diverse amphiphiles exhibit biphasic modulation of GABAA receptors: similarities and differences with neurosteroid actions.
British journal of pharmacology (2010-04-24)
M Chisari, H-J Shu, A Taylor, J H Steinbach, C F Zorumski, Steven Mennerick
ABSTRACT
Some neurosteroids, notably 3alpha-hydroxysteroids, positively modulate GABA(A) receptors, but sulphated steroids negatively modulate these receptors. Recently, other lipophilic amphiphiles have been suggested to positively modulate GABA receptors. We examined whether there was similarity among the actions of these agents and the mechanisms of neurosteroids. Significant similarity would affect theories about the specificity of steroid actions. Xenopus laevis oocytes were challenged with Triton X-100, octyl-beta-glucoside, capsaicin, docosahexaenoic acid and sodium dodecyl sulphate (SDS), along with different GABA concentrations. These compounds have both positive and negative effects on GABA currents, which can be accentuated according to the degree of receptor activation. A low GABA concentration (1 microM) promoted potentiation and a high concentration (20 microM) promoted inhibition of current, except for SDS that inhibited function even at low GABA concentrations. Amphiphile inhibition was characterized by enhanced apparent desensitization and by weak voltage dependence, similar to pregnenolone sulphate antagonism. We then tested amphiphile effects on mutated receptor subunits that are insensitive to negative (alpha1V256S) and positive (alpha1Q241L or alpha1N407A/Y410F) steroid modulation. Negative regulation by amphiphiles was nearly abolished in alpha1V256S-mutated receptors, but potentiation was unaffected. In alpha1Q241L- or alpha1N407A/Y410F-mutated receptors, potentiation by amphiphiles remained intact. Structurally diverse amphiphiles have antagonist actions at GABA(A) receptors very similar to those of sulphated neurosteroids, while the potentiating mechanisms of these amphiphiles are distinct from those of neurosteroid-positive modulators. Thus, such antagonism at GABA(A) receptors does not have a clear pharmacophore requirement.
Sigma-Aldrich
Octyl-β-D-glucopyranoside 100 mM solution
Sigma-Aldrich
Octyl β-D-glucopyranoside solution, ≥95% (HPLC), 50 % (w/v) in H2O
Sigma-Aldrich
Octyl β-D-glucopyranoside, BioXtra, ≥98% (GC)
Sigma-Aldrich
Octyl β-D-glucopyranoside, ≥98% (GC)